Learn More
Sino Biological Recombinant Human ITIH3 Protein (hFc Tag)
Supplier:  Sino Biological 16138H02H100
Encompass_Preferred
A DNA sequence encoding the human (NP_002208.3) (Met1-Asp651) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9933