Learn More
Sino Biological Recombinant Human B7-H7/HHLA2 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 16139H02H50
Encompass_Preferred
A DNA sequence encoding the human HHLA2 (NP_009003.1) (Met1-Asn344) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9938