Learn More
Sino Biological Recombinant Human B7-H7/HHLA2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 16139H08H50
Encompass_Preferred
A DNA sequence encoding the human HHLA2 (NP_009003.1) (Met1-Asn344) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9940