Learn More
Sino Biological Recombinant Human B7-H6/NCR3LG1 Protein (hFc Tag)
Supplier:  Sino Biological 16140H02H200
Encompass_Preferred
A DNA sequence encoding the human NCR3LG1 (NP_001189368.1) (Met1-Ser262) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9941