Learn More
Sino Biological Recombinant Human MTH1 Protein (His Tag)
Supplier:  Sino Biological 16478H07E100
Encompass_Preferred
A DNA sequence encoding the human NUDT1 (NP_002443.3) (Met1-Val156) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9949