Learn More
Sino Biological Recombinant Human FGFR2 Protein (alpha IIIb, hFc Tag), HPLC-verified
Supplier:  Sino Biological 16485H02H200
Encompass_Preferred
A DNA sequence encoding the human FGFR2(alpha(IIIb)) (NP_001138385.1) (Met1-Glu378) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9953