Learn More
Sino Biological Recombinant Human FGFR3 Protein (alpha IIIb, hFc Tag)
Supplier:  Sino Biological 16486H02H200
Encompass_Preferred
A DNA sequence encoding the human FGFR3(alpha(IIIb)) (NP_001156685.1) (Met1-Gly377) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9957