Learn More
Sino Biological Recombinant Human TM2D1 Protein (mFc Tag)
Supplier:  Sino Biological 16487H05H100
Encompass_Preferred
A DNA sequence encoding the human TM2D1 (NP_114416.1) (Met1-Gly118) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-9961