Learn More
Sino Biological Recombinant Human LAG-3 Protein (ECD), Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 16498HNAHB20
Encompass_Preferred
A DNA sequence encoding the Human LAG3 (P18627) (Met1-Arg440) was expressed. The expressed protein was biotinylated in vitro.
Catalog No. 50-161-9973