Learn More
Sino Biological Recombinant Human Niemann Pick C1/NPC1 Protein (His & FLAG Tag), HPLC-verified
Supplier:  Sino Biological 16499H32H100
Encompass_Preferred
A DNA sequence encoding the human NPC1 (NP_000262.2) (Arg372-Phe622) was expressed with a N-terminal polyhistide-tagged FLAG tag at the N-terminus (his-FLAG).
Catalog No. 50-161-9974