Learn More
Sino Biological Recombinant Human TFF3 Protein (His Tag)
Catalog No. 501619977
Encompass_Preferred
A DNA sequence encoding the human TFF3 (NP_003217.3) (Met1-Phe94) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9977 Supplier Sino Biological Supplier No. 16507H08H50
Add to Cart
Add to Cart
A DNA sequence encoding the human TFF3 (NP_003217.3) (Met1-Phe94) was expressed with a polyhistidine tag at the C-terminus.