Learn More
Sino Biological Recombinant Human ILT3/LILRB4 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 16742H02H100
Encompass_Preferred
A DNA sequence encoding the human LILRB4 (NP_001265356.2) (Met1-His257) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9978