Learn More
Cayman Chemical 16α-hydroxy Estrone, Purity: ≥98%, CAS: 566-76-7, 10 mg
SDP
Catalog No. 502972705
Encompass_Preferred
A hydroxylated metabolite of estrone as well as an interconversion product with estriol; formed by CYP1A1, CYP3A5, CYP3A4, and CYP3A7; binds covalently and persistently activates estrogen receptors; increases cell proliferation and does not suppress TNF-α secretion
Catalog No. 50-297-2705 Supplier Cayman Chemical Supplier No. 1520810 MG
A hydroxylated metabolite of estrone as well as an interconversion product with estriol; formed by CYP1A1, CYP3A5, CYP3A4, and CYP3A7; binds covalently and persistently activates estrogen receptors; increases cell proliferation and does not suppress TNF-α secretion