Learn More
Medchemexpress LLC 16a-Hydroxydehydroisoandrosterone | 1232-73-1 | 98.4% | 304.42 | 1 MG
SDP
Catalog No. 5000439738
Encompass_Preferred
16a-Hydroxydehydroisoandrosterone (16α-Hydroxy-DHEA) is a metabolite of the endogenous steroid hormone dehydroepiandrosterone. It is formed from dehydroepiandrosterone via 16-hydroxylation by the cytochrome P450 (CYP) isoforms CYP3A4 and CYP3A5 in adult human liver microsomes, as well as by fetal recombinant CYP3A7. It serves as a precursor to fetal estrogens, including estriol.
  • Metabolite of dehydroepiandrosterone
  • Formed via 16-hydroxylation by CYP3A4, CYP3A5, and CYP3A7 isoforms
  • Precursor to fetal estrogens like estriol
  • For research use only
Catalog No. 50-004-39738 Supplier Medchemexpress LLC Supplier No. HY1130311MG
16a-Hydroxydehydroisoandrosterone (16α-Hydroxy-DHEA) is a metabolite of the endogenous steroid hormone dehydroepiandrosterone. It is formed from dehydroepiandrosterone via 16-hydroxylation by the cytochrome P450 (CYP) isoforms CYP3A4 and CYP3A5 in adult human liver microsomes, as well as by fetal recombinant CYP3A7. It serves as a precursor to fetal estrogens, including estriol.
  • Metabolite of dehydroepiandrosterone
  • Formed via 16-hydroxylation by CYP3A4, CYP3A5, and CYP3A7 isoforms
  • Precursor to fetal estrogens like estriol
  • For research use only