Learn More
TARGETMOL CHEMICALS INC 16alpha-Bromoepiandrosterone
SDP
Catalog No. 502876802
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. 16alpha-Bromoepiandrosterone (BEA) is a water-soluble synthetic sterol related to DHEA and is used in studies of type II diabetes and non-diabetic tuberculosis infections. Purity 98.99%
Catalog No. 50-287-6802 Supplier TARGETMOL CHEMICALS INC Supplier No. T320511MG
Also available in 5 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. 16alpha-Bromoepiandrosterone (BEA) is a water-soluble synthetic sterol related to DHEA and is used in studies of type II diabetes and non-diabetic tuberculosis infections. Purity 98.99%