Learn More
MSC Permatex. 81173 11 oz Cartridge Black RTV Silicone Gasket Sealant -75 to 450°F Operating Temp, 24 hr Full Cure Time
Catalog No. 501296611
Encompass
Permatex. 81173 11 oz Cartridge Black RTV Silicone Gasket Sealant -75 to 450°F Operating Temp, 24 hr Full Cure Time
Catalog No. 50-129-6611 Supplier MSC Supplier No. 85635001
Permatex. 81173 11 oz Cartridge Black RTV Silicone Gasket Sealant -75 to 450°F Operating Temp, 24 hr Full Cure Time