Learn More
STEMCELL Technologies Blunt-End Needles, 16 Gauge, Size: 100 Units
SDP
Catalog No. NC9663175
Encompass
Sterile 16 gauge needles, 1.5 inches long with a metal luer lock hub are ideal for safe and accurate handling of methylcellulose-based medium (e.g. MethoCult™). The blunt end reduces the risk of needle-stick injuries. Product is manufactured from non-pyrogenic materials.
Catalog No. NC9663175 Supplier STEMCELL Technologies Supplier No. 28110
Sterile 16 gauge needles, 1.5 inches long with a metal luer lock hub are ideal for safe and accurate handling of methylcellulose-based medium (e.g. MethoCult™). The blunt end reduces the risk of needle-stick injuries. Product is manufactured from non-pyrogenic materials.