Learn More
International Medical Industries (IMI) Aspirating Needle Blunt Cannula 16 Gauge X 3.5 Inch (Box of 100)
Catalog No. NC1977063
Encompass
Rx-Tract™ 16 Gauge, 3.5” (8.89cm), Blunt Needle provides high-quality solutions for introducing fluids to deep wells and vials, drawing liquids for reconstruction, or pre-fill syringes prior to administration. Rx-Tract Aspirating Needles comes with a conveniently sealed guard to enhance pharmacist and clinician safety. The blunt end prevents accidental needle sticks. •Stainless Steel Needle •Packaged Sterile Non-Pyrogenic •Individual Blister •Box of 100
This item is not returnable. View return policy
Catalog No. NC1977063 Supplier International Medical Industries (IMI) Supplier No. 3234
Rx-Tract™ 16 Gauge, 3.5” (8.89cm), Blunt Needle provides high-quality solutions for introducing fluids to deep wells and vials, drawing liquids for reconstruction, or pre-fill syringes prior to administration. Rx-Tract Aspirating Needles comes with a conveniently sealed guard to enhance pharmacist and clinician safety. The blunt end prevents accidental needle sticks. •Stainless Steel Needle •Packaged Sterile Non-Pyrogenic •Individual Blister •Box of 100