Learn More
Uline Impulse Bag Sealer
Catalog No. NC9124923
Encompass
Impulse sealer, tabletop, 16", replaceable heating element, 8 mil maximum seal thickness (metalized food bags up to 2.5 mil)
Catalog No. NC9124923 Supplier Uline Supplier No. H306
Impulse sealer, tabletop, 16", replaceable heating element, 8 mil maximum seal thickness (metalized food bags up to 2.5 mil)