Learn More
BGB ANALYTIK USA LLC La-Pha-Pack 16ml Screw Neck Vial, 18-400 thread, 71 x 20.6mm, clear glass, 1st hydrolytic class
Supplier:  BGB ANALYTIK USA LLC LP18091704
Encompass
La-Pha-Pack 16ml Screw Neck Vial, 18-400 thread, 71 x 20.6mm, clear glass, 1st hydrolytic class (# 18 09 1704), pk.1000
Catalog No. NC2956047