Learn More
Beckman Coulter Adapter, Sample Cup, 50 mL
Catalog No. NC0888537
Encompass_Preferred
The Beckman Coulter Microfuge 18 Centrifuge Tube Adapter, catalog number 359150, is designed to adapt 0.5mL and 1.5mL microcentrifuge tubes to the Microfuge 18 Centrifuge rotor. It allows centrifugation of up to 6 microcentrifuge tubes at a time. The adapter is made of durable polypropylene and can withstand centrifugal forces up to 18,000 x g. It fits securely into the Microfuge 18 rotor and works with most major brands of microcentrifuge tubes. This adapter provides a convenient way to spin down microsamples in the Microfuge 18 Centrifuge.
Catalog No. NC0888537 Supplier Beckman Coulter Supplier No. 359150
The Beckman Coulter Microfuge 18 Centrifuge Tube Adapter, catalog number 359150, is designed to adapt 0.5mL and 1.5mL microcentrifuge tubes to the Microfuge 18 Centrifuge rotor. It allows centrifugation of up to 6 microcentrifuge tubes at a time. The adapter is made of durable polypropylene and can withstand centrifugal forces up to 18,000 x g. It fits securely into the Microfuge 18 rotor and works with most major brands of microcentrifuge tubes. This adapter provides a convenient way to spin down microsamples in the Microfuge 18 Centrifuge.