Learn More
Ace Glass, Inc. 16MM GRADED SEALS 16MM GRADED SEALS
SDP
Supplier:  Ace Glass, Inc. 845526
Encompass
16mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-598