Learn More
Medicus Health PIERCEABLE TEST TUBE STOPPERS, 16MM, BLUE
SDP
Catalog No. NC2027708
Encompass
∙ 16mm Pierceable Santoprene Stoppers for Vacutainer Test Tubes are available in 5 Colors: Blue, Green, Red, White & Yellow ∙1000 per pack∙Pierceable caps are great for recapping vacuum blood tubes and test tubes∙Caps are produced from santoprene (a rubberized material) and can easily be pierced with a syringe
Catalog No. NC2027708 Supplier Medicus Health Supplier No. 2780M2BLUE
∙ 16mm Pierceable Santoprene Stoppers for Vacutainer Test Tubes are available in 5 Colors: Blue, Green, Red, White & Yellow ∙1000 per pack∙Pierceable caps are great for recapping vacuum blood tubes and test tubes∙Caps are produced from santoprene (a rubberized material) and can easily be pierced with a syringe