Learn More
ALPAQUA ENGINEERING LLC 16MM TUBE RACK
Catalog No. NC0295784
Encompass
The 24-position 16mm tube rack from Alpaqua is an SBS compliant deck accessory for tubes with a diameter of ~15 - 16.6 mm and a length of 70 mm or more. It is compatible with 15 ml conical bottom centrifuge tubes.
Catalog No. NC0295784 Supplier ALPAQUA ENGINEERING LLC Supplier No. A000260
The 24-position 16mm tube rack from Alpaqua is an SBS compliant deck accessory for tubes with a diameter of ~15 - 16.6 mm and a length of 70 mm or more. It is compatible with 15 ml conical bottom centrifuge tubes.