Learn More
MSC CRC Dry Graphite Lube, Dry Film, High Temperature, 10 oz
Catalog No. 501112966
Encompass
CRC Dry Graphite Lube is formulated for dry, high-temperature, non-petroleum lubrication on metal and rubber surfaces. It creates a strong, dry, non-petroleum layer that resists water, repels dust, and can withstand up to 25,000 psi and 850°F intermittently.
  • Dry film lubricant, ideal where petroleum products should be avoided
  • Withstands high temperatures and extreme pressures, as well as water and chemical attack
  • Dries rapidly at room temperature
  • Lowers friction and prevents galling
  • Resists dirt, dust, and oil build-up
  • Non-tacky solid film that does not attract dust, grit, or fibers
  • Will not varnish like multi-purpose lubricants
  • Handles much higher pressure and works in extremely high temperatures
  • Minimal migration once cured
Catalog No. 50-111-2966 Supplier MSC Supplier No. 67964270
CRC Dry Graphite Lube is formulated for dry, high-temperature, non-petroleum lubrication on metal and rubber surfaces. It creates a strong, dry, non-petroleum layer that resists water, repels dust, and can withstand up to 25,000 psi and 850°F intermittently.
  • Dry film lubricant, ideal where petroleum products should be avoided
  • Withstands high temperatures and extreme pressures, as well as water and chemical attack
  • Dries rapidly at room temperature
  • Lowers friction and prevents galling
  • Resists dirt, dust, and oil build-up
  • Non-tacky solid film that does not attract dust, grit, or fibers
  • Will not varnish like multi-purpose lubricants
  • Handles much higher pressure and works in extremely high temperatures
  • Minimal migration once cured