Learn More
MSC CRC Food Grade Chain Lube, Aerosol, Synthetic Blend, 12 oz
Catalog No. 501112978
Encompass
CRC Food Grade Chain Lube, a blend of synthetic lubricants and mineral oil, provides superior lubrication for roller drives and conveyor chains. It penetrates deep to reduce friction and wear, ensuring optimal protection. NSF H1 rated for incidental food contact, suitable for food processing environments.
  • NSF H1 registered for incidental food contact.
  • Perma-Lock® 2-way integrated actuator for broad and precision spray.
  • Non-drying film effectively lubricates metal surfaces.
  • Anti-wear properties extend chain life.
  • Foaming action ensures deep penetration without messy overspray.
  • Reduces friction and wear, extending equipment life.
Catalog No. 50-111-2978 Supplier MSC Supplier No. 91990978
CRC Food Grade Chain Lube, a blend of synthetic lubricants and mineral oil, provides superior lubrication for roller drives and conveyor chains. It penetrates deep to reduce friction and wear, ensuring optimal protection. NSF H1 rated for incidental food contact, suitable for food processing environments.
  • NSF H1 registered for incidental food contact.
  • Perma-Lock® 2-way integrated actuator for broad and precision spray.
  • Non-drying film effectively lubricates metal surfaces.
  • Anti-wear properties extend chain life.
  • Foaming action ensures deep penetration without messy overspray.
  • Reduces friction and wear, extending equipment life.