Learn More
C&G Containers 16OZ Amber Glass Boston Round Bottle, UV Protection, High Chemical Resistance, 12 Bottles
SDP
Catalog No. NC0864734
Encompass_Preferred
The 16OZ AMBER BOTTLE is a versatile solution for safely storing light-sensitive substances. Crafted from durable glass, this Boston Round bottle boasts high chemical resistance and essential UV protection thanks to its amber coloration. With a 28-400 neck finish, it's compatible with standard closures, ensuring secure storage for laboratory and pharmaceutical applications. Supplied in a case of 12, it allows for efficient inventory management, making it an ideal choice for a range of uses.
Catalog No. NC0864734 Supplier C&G Containers Supplier No. LAS016200010
The 16OZ AMBER BOTTLE is a versatile solution for safely storing light-sensitive substances. Crafted from durable glass, this Boston Round bottle boasts high chemical resistance and essential UV protection thanks to its amber coloration. With a 28-400 neck finish, it's compatible with standard closures, ensuring secure storage for laboratory and pharmaceutical applications. Supplied in a case of 12, it allows for efficient inventory management, making it an ideal choice for a range of uses.