Learn More
MSC Bernzomatic Propane Tank, 16 oz, Steel, Disposable, 1 ea
Catalog No. NC2884123
Encompass
The all-purpose propane gas cylinder provides 16 oz of clean-burning, portable propane fuel. It is constructed with lightweight, durable steel and features reliable valves for safe and easy use.
  • Provides 16 oz of clean-burning, portable propane fuel
  • Ideal for on-the-go cooking, grilling, heating, and lighting
  • Constructed with lightweight, durable steel
  • Features reliable valves for safe and easy use
  • Compatible with standard portable propane appliances and torches with a CGA 600 connection
  • Manufactured to exceed government agency requirements
  • Pre-filled
This item is not returnable. View return policy
Catalog No. NC2884123 Supplier MSC Supplier No. 70132618
The all-purpose propane gas cylinder provides 16 oz of clean-burning, portable propane fuel. It is constructed with lightweight, durable steel and features reliable valves for safe and easy use.
  • Provides 16 oz of clean-burning, portable propane fuel
  • Ideal for on-the-go cooking, grilling, heating, and lighting
  • Constructed with lightweight, durable steel
  • Features reliable valves for safe and easy use
  • Compatible with standard portable propane appliances and torches with a CGA 600 connection
  • Manufactured to exceed government agency requirements
  • Pre-filled