Learn More
US Plastic Corporation 16 oz. HDPE White Canister with 89mm Neck (Lid Sold Separately)
SDP
Catalog No. NC1521285
Encompass
White opaque canisters are made from high density polyethylene and molded white in color. Ideal for most food products as well as chemicals, cosmetics and powders.
Catalog No. NC1521285 Supplier US Plastic Corporation Supplier No. 2579
White opaque canisters are made from high density polyethylene and molded white in color. Ideal for most food products as well as chemicals, cosmetics and powders.