Learn More
Essendant Microwavable Deli Containers, 16 oz, Clear, 500/Carton
Catalog No. NC1395384
Encompass_Preferred
Package salads, soups and more in flavor- and odor-transfer-proof containers. Printable surface provides branding and labeling opportunities. Smooth-rolled, doubled-over rims enhance rigidity. Product line's standardized diameter delivers one-lid-fits-all convenience. Microwave-safe polyethylene withstands temperatures of up to 240¿F.
Catalog No. NC1395384 Supplier Essendant Supplier No. FABPK16SC
Package salads, soups and more in flavor- and odor-transfer-proof containers. Printable surface provides branding and labeling opportunities. Smooth-rolled, doubled-over rims enhance rigidity. Product line's standardized diameter delivers one-lid-fits-all convenience. Microwave-safe polyethylene withstands temperatures of up to 240¿F.