Learn More
Cayman Chemical 16phenoxy ttrnor PROSGla 10mg
SDP
Catalog No. 502967948
Encompass_Preferred
16-phenoxy tertanor PGF2α is a metabolically stable analog of PGF2α. It binds to the FP receptor on ovine luteal cells with much greater affinity (440%) than PGF2α.{2058}
Catalog No. 50-296-7948 Supplier Cayman Chemical Supplier No. 1676010 MG
16-phenoxy tertanor PGF2α is a metabolically stable analog of PGF2α. It binds to the FP receptor on ovine luteal cells with much greater affinity (440%) than PGF2α.{2058}