Learn More
Perkin Elmer US LLC 16-Position Turntable with Standard 75 mL, 40 Bar Digestion Vessels, without DPC, Complete Assembly
Catalog No. 502111677
Encompass_Preferred
This assembly includes: 16 Standard 75 mL (40 Bar) Digestion Vessels made of TFM and lids Rupture Disc Vessel cap (without DPC) 16-Position Turntable Gas Containment Manifold
Catalog No. 50-211-1677 Supplier Perkin Elmer US LLC Supplier No. N3131005
Add to Cart
Add to Cart
Request a Quote
This assembly includes: 16 Standard 75 mL (40 Bar) Digestion Vessels made of TFM and lids Rupture Disc Vessel cap (without DPC) 16-Position Turntable Gas Containment Manifold