Learn More
Kuhner Shaker Inc 16X WHITE POLYSTYRENE SQUARE H
Catalog No. 502570136
Encompass
16x white polystyrene square half deepwell MTP, transparent polystyrene bottom, 96 wells of 1.1 ml volume, gamma-irradiated)
Catalog No. 50-257-0136 Supplier Kuhner Shaker Inc Supplier No. SMCR1496D
16x white polystyrene square half deepwell MTP, transparent polystyrene bottom, 96 wells of 1.1 ml volume, gamma-irradiated)