Learn More
Sarstedt Inc 16X100 TUBE 13ML RB PP/PK1000
Catalog No. NC0548330 Shop All Sarstedt Inc. Products
Encompass_Preferred
13ml Tube 16.8x100mm Round Bottom Polypropylene No Cap
Catalog No. NC0548330 Supplier Sarstedt Inc Supplier No. 55.515
13ml Tube 16.8x100mm Round Bottom Polypropylene No Cap