Learn More
LAKE CHARLES MFG 16x100mm Test Tubes, Polypropylene, Round BTM, Natural, 14ml Vol, Karter Scientific 206U2 (Pack of 250)
Catalog No. NC1520311
Encompass
Cylindrical test tubes made of PP material. Molded from premium food and laboratory grade polymers. These general purpose tubes are ideal for use in many applications from laboratory analysis to food and beverage service. Our tubes have passed a rigorous inspection by our Quality Assurance Department to ensure they exceed our exacting specifications. Polypropylene tubes are autoclavable, dishwasher safe, chemically resistant, and virtually unbreakable. Although not as clear as Polystyrene, Polypropylene is durable and does not scratch easily.
Catalog No. NC1520311 Supplier LAKE CHARLES MFG Supplier No. 206U2
Cylindrical test tubes made of PP material. Molded from premium food and laboratory grade polymers. These general purpose tubes are ideal for use in many applications from laboratory analysis to food and beverage service. Our tubes have passed a rigorous inspection by our Quality Assurance Department to ensure they exceed our exacting specifications. Polypropylene tubes are autoclavable, dishwasher safe, chemically resistant, and virtually unbreakable. Although not as clear as Polystyrene, Polypropylene is durable and does not scratch easily.