Learn More
LAKE CHARLES MFG 16x150mm Test Tubes, Polystyrene, Round BTM, 20ml Vol, Clear, Karter Scientific 400A1 (Case of 1000)
Catalog No. NC2180892
Encompass
Cylindrical test tube, manufactured from high quality polystyrene. PS Tubes are not autoclavable or dishwasher safe but are chemically resistant and clear like glass. Polystyrene has the properties of glass but at a much lower price point. Since polystyrene is not as hard as glass it will have minor scratches. Colors is clear
This item is not returnable. View return policy
Catalog No. NC2180892 Supplier LAKE CHARLES MFG Supplier No. 400A1
Cylindrical test tube, manufactured from high quality polystyrene. PS Tubes are not autoclavable or dishwasher safe but are chemically resistant and clear like glass. Polystyrene has the properties of glass but at a much lower price point. Since polystyrene is not as hard as glass it will have minor scratches. Colors is clear