Learn More
Uline 2 Mil Industrial Poly Bags, Clear, 1000 EA
Catalog No. NC2106386
Encompass_Preferred
Most popular thickness for shipping parts, clothing or protecting food. 100% virgin high clarity polyethylene film. Meets FDA and USDA specifications for food contact.
Catalog No. NC2106386 Supplier Uline Supplier No. S1352
Most popular thickness for shipping parts, clothing or protecting food. 100% virgin high clarity polyethylene film. Meets FDA and USDA specifications for food contact.