Learn More
MSC Value Collection Air Filter, MERV 8, 16x20x1 in, 1 ea
Catalog No. 5000504011
Encompass
This pleated air filter is designed for efficient filtration in Heating & Air Conditioning Units. Featuring a MERV 8 rating and synthetic media, it provides effective particle capture for improved air quality.
  • Measures 16 in high, 20 in wide, and 1 in deep
  • Constructed with a sturdy moisture resistant beverage board frame
  • Utilizes exclusive vForm Self-Supported Pleats
  • Operates with an air flow of 670 CFM and a maximum velocity of 300 FPM
  • Meets ASHRAE 52.2-2012 and UL 900 standards
  • Withstands maximum temperatures of 150°F
  • Initial resistance of 0.2900 in wg
Catalog No. 50-005-04011 Supplier MSC Supplier No. 03639614
This pleated air filter is designed for efficient filtration in Heating & Air Conditioning Units. Featuring a MERV 8 rating and synthetic media, it provides effective particle capture for improved air quality.
  • Measures 16 in high, 20 in wide, and 1 in deep
  • Constructed with a sturdy moisture resistant beverage board frame
  • Utilizes exclusive vForm Self-Supported Pleats
  • Operates with an air flow of 670 CFM and a maximum velocity of 300 FPM
  • Meets ASHRAE 52.2-2012 and UL 900 standards
  • Withstands maximum temperatures of 150°F
  • Initial resistance of 0.2900 in wg