Learn More
Medchemexpress LLC 17:0-17:1-17:0 TG-d5 | 958760-74-2 | 99.9% | 852.41 | 1 MG
SDP
Catalog No. 5000363637
Encompass_Preferred
17:0-17:1-17:0 TG-d5 is a deuterium-labeled derivative of 17:0-17:1-17:0 TG, designed for research applications. This compound is primarily utilized as a tracer and internal standard in quantitative analytical techniques.
  • Used as a tracer in research studies.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes to influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-63637 Supplier Medchemexpress LLC Supplier No. HY146915S1MG
Add to Cart
Add to Cart
17:0-17:1-17:0 TG-d5 is a deuterium-labeled derivative of 17:0-17:1-17:0 TG, designed for research applications. This compound is primarily utilized as a tracer and internal standard in quantitative analytical techniques.
  • Used as a tracer in research studies.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes to influence pharmacokinetic and metabolic profiles of drugs.