Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 17:1 lyso pc | 1246304-62-0 | 98.0% | 507.64 | C25H50NO7P | 10 MG
SDP

Catalog No. 5000286907
Encompass_Preferred

17:1 Lyso PC is a lysophosphatidylcholine lipid used to simulate biological phospholipid membranes and to formulate liposomes for drug delivery research. It forms vesicles with concentric phospholipid bilayers, enabling encapsulation of polar, water-soluble payloads in the internal aqueous space and incorporation of lipophilic payloads within the lipid layer. It is commonly used in studies of anticancer, anti-infection, and nucleic-acid delivery.

  • Molecular formula: C25H50NO7P.
  • Molecular weight: 507.64.
  • Purity: 98.0%.
  • CAS number: 1246304-62-0.
  • Appearance: white to off-white solid-liquid mixture.
  • Storage: -20°C (pure form, long term); -80°C (in solvent, short term).
  • Pack size: 10 mg.
  • Typical applications: liposome formulation and nucleic-acid delivery research.

Catalog No. 50-002-86907 Supplier Medchemexpress LLC Supplier No. HY15372510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

17:1 Lyso PC is a lysophosphatidylcholine lipid used to simulate biological phospholipid membranes and to formulate liposomes for drug delivery research. It forms vesicles with concentric phospholipid bilayers, enabling encapsulation of polar, water-soluble payloads in the internal aqueous space and incorporation of lipophilic payloads within the lipid layer. It is commonly used in studies of anticancer, anti-infection, and nucleic-acid delivery.

  • Molecular formula: C25H50NO7P.
  • Molecular weight: 507.64.
  • Purity: 98.0%.
  • CAS number: 1246304-62-0.
  • Appearance: white to off-white solid-liquid mixture.
  • Storage: -20°C (pure form, long term); -80°C (in solvent, short term).
  • Pack size: 10 mg.
  • Typical applications: liposome formulation and nucleic-acid delivery research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.