Learn More
Medchemexpress LLC 17:1 Lyso PC | 1246304-62-0 | C25H50NO7P | 25 MG
SDP
Catalog No. 5000375560
Encompass_Preferred
17:1 Lyso PC is a liposome designed to simulate biological phospholipid membranes. These liposomes, comprising concentric phospholipid bilayer membranes, are utilized in drug delivery systems for anti-cancer and anti-infection applications. It effectively encapsulates water-soluble payloads in its aqueous core and integrates lipophilic payloads into its lipid bilayer, proving particularly effective for antisense oligonucleotide delivery by overcoming issues like inefficient cellular uptake and rapid loss in the body.
  • Simulates biological phospholipid membranes
  • Forms concentric phospholipid bilayer vesicles
  • Used in drug delivery systems for anti-cancer and anti-infection fields
  • Encapsulates water-soluble payloads in aqueous core
  • Integrates lipophilic payloads into lipid bilayer
  • Addresses inefficient cellular uptake and rapid loss for antisense oligonucleotide delivery
Catalog No. 50-003-75560 Supplier Medchemexpress LLC Supplier No. 25MGHY153725
17:1 Lyso PC is a liposome designed to simulate biological phospholipid membranes. These liposomes, comprising concentric phospholipid bilayer membranes, are utilized in drug delivery systems for anti-cancer and anti-infection applications. It effectively encapsulates water-soluble payloads in its aqueous core and integrates lipophilic payloads into its lipid bilayer, proving particularly effective for antisense oligonucleotide delivery by overcoming issues like inefficient cellular uptake and rapid loss in the body.
  • Simulates biological phospholipid membranes
  • Forms concentric phospholipid bilayer vesicles
  • Used in drug delivery systems for anti-cancer and anti-infection fields
  • Encapsulates water-soluble payloads in aqueous core
  • Integrates lipophilic payloads into lipid bilayer
  • Addresses inefficient cellular uptake and rapid loss for antisense oligonucleotide delivery