Learn More
Medchemexpress LLC 17:1 Lyso PC | 1246304-62-0 | C25H50NO7P | 5 MG
SDP
Catalog No. 5000376166
Encompass_Preferred
17:1 Lyso PC is a liposome designed to simulate biological phospholipid membranes. Liposomes are the primary component of vesicles with concentric phospholipid bilayer membranes, which can be used to construct drug delivery systems for anti-cancer and anti-infection fields.
  • Encapsulates highly polar water-soluble payloads within the internal aqueous space.
  • Allows lipophilic payloads to partition into and become part of the lipid bilayer.
  • Useful for delivering antisense oligonucleotides.
  • Addresses inefficient cellular uptake and rapid loss in the body.
Catalog No. 50-003-76166 Supplier Medchemexpress LLC Supplier No. HY1537255MG
17:1 Lyso PC is a liposome designed to simulate biological phospholipid membranes. Liposomes are the primary component of vesicles with concentric phospholipid bilayer membranes, which can be used to construct drug delivery systems for anti-cancer and anti-infection fields.
  • Encapsulates highly polar water-soluble payloads within the internal aqueous space.
  • Allows lipophilic payloads to partition into and become part of the lipid bilayer.
  • Useful for delivering antisense oligonucleotides.
  • Addresses inefficient cellular uptake and rapid loss in the body.