Learn More
Avanti Polar Lipids 1-(10Z-heptadecenoyl)-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium salt), 1246298-11-2, MFCD22416584, 5 mg
Catalog No. NC1619534
Encompass
Molecular Formula: C23H44NaO9P, Formula Weight: 518.554, Purity >99% LPG; may contain up to 15% of the 2-LPG isomer.
Catalog No. NC1619534 Supplier Avanti Polar Lipids Supplier No. 858127C5MG
Molecular Formula: C23H44NaO9P, Formula Weight: 518.554, Purity >99% LPG; may contain up to 15% of the 2-LPG isomer.