Learn More
Medchemexpress LLC 17,17-(Ethylenedioxy)androst-4-en-3-one | 1044-89-9 | 99.6% | 330.46 | 1 MG
SDP
Catalog No. 5000344554
Encompass_Preferred
17,17-(Ethylenedioxy)androst-4-en-3-one is an effective ingredient in cosmetics. It can be used for acne and to promote hair growth research. This compound inhibits reductase activity and the bond of a receptor protein and 5α-DHT.
  • Effective ingredient in cosmetics
  • Used for acne research
  • Promotes hair growth research
  • Inhibits reductase activity
  • Inhibits bond of a receptor protein and 5α-DHT
Catalog No. 50-003-44554 Supplier Medchemexpress LLC Supplier No. HY1089525G
17,17-(Ethylenedioxy)androst-4-en-3-one is an effective ingredient in cosmetics. It can be used for acne and to promote hair growth research. This compound inhibits reductase activity and the bond of a receptor protein and 5α-DHT.
  • Effective ingredient in cosmetics
  • Used for acne research
  • Promotes hair growth research
  • Inhibits reductase activity
  • Inhibits bond of a receptor protein and 5α-DHT