Learn More
Medchemexpress LLC 17,17-(Ethylenedioxy)androst-4-en-3-one | 1044-89-9 | 99.6% | 330.46 | 5 G
SDP
Catalog No. 5000345970
Encompass_Preferred
17,17-(Ethylenedioxy)androst-4-en-3-one (also known as 4-androstene-3, 17-dione-17-cyclic ethylene ketal) is an effective ingredient in cosmetics. It can be used for acne and to promote hair growth research. This compound can inhibit reductase activity and also inhibit the bond of a receptor protein and 5α-DHT.
  • Effective ingredient in cosmetics
  • Used for acne and hair growth research
  • Inhibits reductase activity
  • Inhibits the bond of a receptor protein and 5α-DHT
  • Target: 5 alpha reductase
  • Pathway: metabolic enzyme/protease
Catalog No. 50-003-45970 Supplier Medchemexpress LLC Supplier No. HY10895210G
17,17-(Ethylenedioxy)androst-4-en-3-one (also known as 4-androstene-3, 17-dione-17-cyclic ethylene ketal) is an effective ingredient in cosmetics. It can be used for acne and to promote hair growth research. This compound can inhibit reductase activity and also inhibit the bond of a receptor protein and 5α-DHT.
  • Effective ingredient in cosmetics
  • Used for acne and hair growth research
  • Inhibits reductase activity
  • Inhibits the bond of a receptor protein and 5α-DHT
  • Target: 5 alpha reductase
  • Pathway: metabolic enzyme/protease