Learn More
Cayman Chemical 17 18EpETE methyl ester 100ug
SDP
Catalog No. 502953813
Encompass_Preferred
An esterified form of (±)17(18)-EpETE; more lipid soluble than (±)17(18)-EpETE
Catalog No. 50-295-3813 Supplier Cayman Chemical Supplier No. 13367100 UG
An esterified form of (±)17(18)-EpETE; more lipid soluble than (±)17(18)-EpETE