Learn More
MOLDPRO INC 15 mL Digestion Tube w/o cap, 17.5 x 106 mm, 1000/case, Flat Bottom, Graduated, PP Homopolymer, includes COA for Volume and Background Metals
Catalog No. NC1652174
Encompass_Preferred
MoldPro 15 mL Digestion Tubes (MP-123 and MP-123PN) are certified to class A volume accuracy, include a metals background report and are compatible with leading brand graphite block digestion systems• 1,000 vessels/case • Made from virgin polypropylene • Excellent visibility • Very good resistance to acids • Molded-in graduations are in 1 mL increments to 15 mL • 5, 10 and 15mL graduation lines extend around the entire circumference • Total capacity of 18 mL • Extremely low leachable metal content; each shipment comes with a metals background report • Fit leading brand graphite digestion block systems • Can be used as a digestion vessel, graduated cylinder, volumetric flask, autosampler tube, or storage container • We also offer ribbed watch glasses (MP-128) • Our 15 mL digestion tube is a direct replacement for Environmental Express SC415 and SCP Science 010-515-263 and fit the EE SC196 96 well HotBlock and 15ml DigiPREP Mini, Jr and LS systems.
Catalog No. NC1652174 Supplier MOLDPRO INC Supplier No. MP123
MoldPro 15 mL Digestion Tubes (MP-123 and MP-123PN) are certified to class A volume accuracy, include a metals background report and are compatible with leading brand graphite block digestion systems• 1,000 vessels/case • Made from virgin polypropylene • Excellent visibility • Very good resistance to acids • Molded-in graduations are in 1 mL increments to 15 mL • 5, 10 and 15mL graduation lines extend around the entire circumference • Total capacity of 18 mL • Extremely low leachable metal content; each shipment comes with a metals background report • Fit leading brand graphite digestion block systems • Can be used as a digestion vessel, graduated cylinder, volumetric flask, autosampler tube, or storage container • We also offer ribbed watch glasses (MP-128) • Our 15 mL digestion tube is a direct replacement for Environmental Express SC415 and SCP Science 010-515-263 and fit the EE SC196 96 well HotBlock and 15ml DigiPREP Mini, Jr and LS systems.