Learn More
AdipoGen 17-AAG
SDP
Supplier:  AdipoGen BVT0244C100
Encompass_Preferred
Chemical. CAS 75747-14-7. Formula C31H43N3O8. MW 585.7. Semi-synthetic derivative from geldanamycin. Potent, less toxic analog of geldanamycin Prod. No. BVT-0196. Inhibits the essential ATPase activity of HSP90. Telomerase activity inhibitor. Apoptosis inducer. Antitumor compound.
Catalog No. 50-256-2050