Learn More
Alkali Scientific 17 AAG [17(Allyamino) 17 demothoxygeldanamycin], 5 Milligrams
SDP
Supplier:  Alkali Scientific AAG003CP
Encompass_Preferred
17-AAG (17-Allylamino-17-demethoxygeldanamycin) is a potent Hsp90 inhibitor widely used in anticancer and molecular biology research. It is a semi-synthetic analog of geldanamycin that disrupts the function of heat shock protein 90, a chaperone protein essential for stabilizing many oncogenic client proteins. By inhibiting Hsp90, 17-AAG promotes degradation of proteins involved in cancer cell growth, survival, and signaling pathways. This compound is commonly used in studies investigating tumor biology, apoptosis, and targeted cancer therapies. It is suitable for in vitro research applications, including cell culture experiments and biochemical assays focused on protein folding, stress response, and signal transduction mechanisms.
Catalog No. 50-006-60799