Learn More
Apexbio Technology LLC 17-AAG (KOS953) 75747-14-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063970
Encompass_Preferred
17-AAG (KOS953) is a synthetic derivative of geldanamycin that inhibits heat shock protein 90 (HSP90) It acts by binding to HSP90 causing destabilization of client proteins including HER2 Raf-1 p53 and components of MAPK signaling pathways Reported IC50 for 17-AAG in BT474 cells is approximately 6 nM Its biological activity has been investigated broadly including in multiple myeloma breast cancer thyroid cancer Hodgkin lymphoma and melanoma cellular models Ongoing studies explore 17-AAG both as monotherapy and in combination with agents like bortezomib or trastuzumab for addressing treatment-resistant cancer cell lines Currently 17-AAG is evaluated in Phase II clinical trials
Catalog No. 50-000-63970 Supplier Apexbio Technology LLC Supplier No. A40545.1
17-AAG (KOS953) is a synthetic derivative of geldanamycin that inhibits heat shock protein 90 (HSP90) It acts by binding to HSP90 causing destabilization of client proteins including HER2 Raf-1 p53 and components of MAPK signaling pathways Reported IC50 for 17-AAG in BT474 cells is approximately 6 nM Its biological activity has been investigated broadly including in multiple myeloma breast cancer thyroid cancer Hodgkin lymphoma and melanoma cellular models Ongoing studies explore 17-AAG both as monotherapy and in combination with agents like bortezomib or trastuzumab for addressing treatment-resistant cancer cell lines Currently 17-AAG is evaluated in Phase II clinical trials